Learn More
Fisher BioReagents™ PCR Nucleotide Mix
A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR
Brand: Fisher BioReagents™ BP2565-1
Code : 32
Additional Details : CAS Number : 18423-43-3 Weight : 0.05100kg
Description
- Functionally tested in PCR
- High purity: >98% triphosphate
- Supplied as 10mM solution in purified water (pH 7.5)
Specifications
Liquid | |
-20°C | |
>98% | |
Colorless |
7.5 | |
10 mM | |
1000 μL | |
PCR |