Learn More
Brand™ PCR Plate Combo Pack PROMO
Product Code. 15594409 Shop All Brand Products
Click to view available options
Description:
Elevated rim low profile plates
Non-Skirted, Low profile plates
Non-skirted PCR plates
Semi-Skirted, Elevated, Low profile plates
Standard profile plates
Unit Size:
1 set
Product Code. 15594409
Brand: Brand™ 781230
Includes
96-well PCR plates, PCR-sealing film
Provides additional savings than when plates and fiilm are purchased individually. Brand™ PCR Plate Combo Pack PROMO includes 96-well PCR plates available in different skirt and profile types along with high quality qPCR sealing film.
Premium PCR/qPCR transparent plates
  • 96-well
  • Skirted, non-skirted, or semi-skirted
  • Low, standard or elevated rim profile
  • Ultra-thin walled for quick and constant thermal transfer
  • Free of DNase, DNA, RNase and endotoxins
  • No warping, no wiggling, suitable for automation
Includes qPCR PCR sealing film:
  • Highly transparent for qPCR
  • Press to seal
  • Convenient end tabs
  • No annoying adhesive residues
TRUSTED_SUSTAINABILITY
Specifications
Description Non-Skirted, Low profile plates
Quantity 50/Pk.
No. of Wells 96
Format 96-well
Product Type PCR Combo Pack