Learn More
Invitrogen™ PCQAP Monoclonal Antibody (6F4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549257
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human U87 whole cell, human CACO-2 whole cell, human HL-60 whole cell, human HepG2 whole cell, human Hela whole cell, human PC-3 whole cell. ICC/IF: A431 cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a transcription factor that represses GATA-regulated genes and binds to a dynein light chain protein. Binding of the encoded protein to the dynein light chain protein affects binding to GATA consensus sequences and suppresses its transcriptional activity. Defects in this gene are a cause of tricho-rhino-phalangeal syndrome (TRPS) types I-III.
Specifications
| PCQAP | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| Q96RN5 | |
| MED15 | |
| E. coli-derived human MED15 recombinant protein (Position: M1-A285). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 6F4 | |
| Unconjugated | |
| MED15 | |
| A230074L19Rik; Activator-recruited cofactor 105 kDa component; activator-recruited cofactor, 105-kD; ARC105; AW536074; CAG7A; CTG repeat protein 7a; CTG7A; DKFZp686A2214; DKFZp762B1216; FLJ42282; FLJ42935; MED15; Mediator complex subunit 15; mediator of RNA polymerase II transcription subunit 15; mPcqap; PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associated protein; PC2 glutamine/Q-rich-associated protein; PC2-glutamine-rich-associated protein; PCQAP; Positive cofactor 2 glutamine/Q-rich-associated protein; positive cofactor 2, glutamine/Q-rich-associated protein; positive cofactor 2, multiprotein complex, glutamine/Q-rich-associated protein; TIG1; TIG-1; TNRC7; TPA inducible gene-1; TPA inducible protein; TPA-inducible gene 1 protein; trinucleotide repeat containing 7; trinucleotide repeat-containing gene 7 protein | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 51586 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.