missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCPTP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529243
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299. Predicted reactivity: Mouse (96%), Rat (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and a single intracellular catalytic domains, and thus represents a receptor-type PTP. The similar gene predominately expressed in mouse brain was found to associate with, and thus regulate the activity and cellular localization of MAP kinases. The rat counterpart of this gene was reported to be regulated by the nerve growth factor, which suggested the function of this gene in neuronal growth and differentiation.
Specifications
| PCPTP1 | |
| Polyclonal | |
| Unconjugated | |
| PTPRR | |
| Ch-1 PTPase; ch-1PTPase; ECPPT; ECPTP; EC-PTP; Gmcp1; mPTP213; NC-PTPCOM1; PC12-PTP1; PCPTP1; Phosphotyrosine phosphatase 13; protein tyrosine phosphatase Cr1PTPase; protein tyrosine phosphatase PCPTP1; protein tyrosine phosphatase, receptor type R; protein tyrosine phosphatase, receptor type, R; protein-tyrosine phosphatase NC-PTPCOM1; protein-tyrosine phosphatase PCPTP1; Protein-tyrosine-phosphatase SL; protein-tyrosine-phosphatase-SL; Ptp; Ptp13; PTPBR7; PTPRQ; PTPRR; PTP-SL; receptor-type tyrosine-protein phosphatase R; R-PTP-R; RPTPRR; STEP-like PTPase; tyrosine phosphatase CBPTP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.56 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q15256 | |
| PTPRR | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human PCPTP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction