missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCP2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549455
This item is not returnable.
View return policy
Description
May function as a cell-type specific modulator for G protein-mediated cell signaling.
Specifications
| PCP2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| Q8IVA1 | |
| PCP2 | |
| KLH conjugated synthetic peptide between 91-125 amino acids from the Central region of human PCP2. | |
| 80 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PCP2 | |
| Purkinje cell protein 2 homolog, PCP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 126006 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction