missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCOLCE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112643
This item is not returnable.
View return policy
Description
PCOLCE2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Binds to the C-terminal propeptide of types I and II procollagens and may enhance the cleavage of that propeptide by BMP1.
Specifications
| PCOLCE2 | |
| Polyclonal | |
| Unconjugated | |
| PCOLCE2 | |
| 2400001O18Rik; PCOLCE2; PCPE2; PCPE-2; Procollagen C-endopeptidase enhancer 2; procollagen COOH-terminal proteinase enhancer 2; procollagen C-proteinase enhancer 2; UNQ250/PRO287 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26577 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UKZ9 | |
| PCOLCE2 | |
| Recombinant Human Procollagen C-endopeptidase enhancer 2 protein (24-415AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction