missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCNXL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553164
This item is not returnable.
View return policy
Description
Immunogen sequence: LPLHQEVDSS DSEVAVTLID TSQPGDPLSL HEPIKIVITM SSTPNSMTDL ESSLHLRVVG TEKTSVKSDA EPTNPGAAGS PNAEQISIPV ITLDLPEGGG GGVPCPE Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 63%.
May play a role in tumorigenesis of colorectal carcinomas with high microsatellite instability (MSI-H).
Specifications
| PCNXL2 | |
| Polyclonal | |
| Unconjugated | |
| PCNX2 | |
| A830048P05; AU067631; E330039K12Rik; KIAA0435; mKIAA0435; PCNX2; Pcnxl2; pecanex 1; pecanex homolog 2 (Drosophila); Pecanex homolog protein 2; pecanex-like 2 (Drosophila); pecanex-like protein 2; RGD1305883 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NKB5 | |
| PCNX2 | |
| Recombinant protein corresponding to Human PCNXL2. Recombinant protein control fragment (Product #RP-90604). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction