missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCNX Polyclonal Antibody

Description
Immunogen sequence: VDPSQILEGI NLSKRKELQW PDEGIRLKAG RNSWKDWSPQ EGMEGHVIHR WVPCSRDPGT RSHIDKAVLL VQIDDKYVTV IETGVLELGA EV.
This gene encodes an evolutionarily conserved transmembrane protein similar to the pecanex protein in Drosophila. The fly protein is a component of the Notch signaling pathway, which functions in several developmental processes.
Specifications
Specifications
| Antigen | PCNX |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PCNX1 |
| Gene Accession No. | Q96RV3 |
| Gene Alias | KIAA0805; KIAA0995; Pcnx; PCNX1; PCNXL1; pecanex; pecanex 1; pecanex homolog (Drosophila); pecanex homolog 1 (Drosophila); Pecanex homolog protein 1; Pecanex-like protein 1 |
| Gene Symbols | PCNX1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?