missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCNP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557113
This item is not returnable.
View return policy
Description
Immunogen sequence: NEDEDSEPEE MPPEAKMRMK NIGRDTPTSA GPNSFNKGKH GFSDNQKLWE RNIKSHLGNV HDQD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PCNP is a protein coding gene, and may be involved in cell cycle regulation.
Specifications
| PCNP | |
| Polyclonal | |
| Unconjugated | |
| PCNP | |
| 1110018D06Rik; Ab2-416; AI647035; Liver regeneration-related protein LRRG084; PCNP; PEST proteolytic signal containing nuclear protein; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein; Unknown (protein for MGC:128940); zgc:103440 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WW12 | |
| PCNP | |
| Recombinant protein corresponding to Human PCNP. Recombinant protein control fragment (Product #RP-96291). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction