missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35562-100ul
This item is not returnable.
View return policy
Description
PCNP Polyclonal antibody specifically detects PCNP in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PCNP | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-178 of human PCNP (NP_065090.1).,, Sequence:, MADGKAGDEKPEKSQRAGAAGGPEEEAEKPVKTKTVSSSNGGESSSRSAEKRSAEEEAADLPTKPTKISKFGFAIGSQTTKKASAISIKLGSSKPKETVPTLAPKTLSVAAAFNEDEDSEPEEMPPEAKMRMKNIGRDTPTSAGPNSFNKGKHGFSDNQKLWERNIKSHLGNVHDQDN | |
| 100 μL | |
| Cell Cycle and Replication | |
| 57092 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction