missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37692
This item is not returnable.
View return policy
Description
PCNP Polyclonal specifically detects PCNP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PCNP | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp781I24156, PEST proteolytic signal containing nuclear protein, PEST proteolytic signal-containing nuclear protein, PEST-containing nuclear protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:10000 - 1:25000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_065090.1 | |
| PCNP | |
| This antibody maps to a region between residue 1 to 50 of human PEST Proteolytic Signal Containing Nuclear Protein using the numbering given in entry NP_065090.1 (GeneID 57092). | |
| 100 μL | |
| Cell Cycle and Replication | |
| 57092 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction