missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCNA Recombinant Rabbit Monoclonal Antibody (144), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£218.00 - £436.00
Specifications
| Antigen | PCNA |
|---|---|
| Clone | 144 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17789773
|
Invitrogen™
MA543430 |
50 μL |
£218.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17729683
![]() |
Invitrogen™
MA543131 |
100 μL |
£436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human PCNA
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.Specifications
| PCNA | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C | |
| Pcna | |
| IgG | |
| Protein A | |
| Antibody |
| 144 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P12004 | |
| 5111 | |
| Recombinant Human PCNA Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Pcna |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title