missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24036-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody specifically react with the 36 kDa human PCNA protein.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein associates with formin-binding protein 17, but its function is unknown. This protein may form oligomeric complexes with family members.Specifications
| PCNA | |
| Polyclonal | |
| Unconjugated | |
| PCNA | |
| Cyclin, PCNA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18538, 25737, 5111 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04961, P12004, P17918 | |
| PCNA | |
| PCNA Fusion Protein Ag7416 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction