missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Shop All Epredia ProductsDescription
Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase delta auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-delta to play a fundamental role in the initiation of cell proliferation.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant human PCNA protein
Molecular Weight: 36kDa
Positive Control: Raji cells, Tonsil, Lymph node, and small intestine
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | PCNA (Proliferating Cell Nuclear Antigen) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant human PCNA protein |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cell Cycle |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?