missing translation for 'onlineSavingsMsg'
Learn More

PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Product Code. 12603427 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
0.1mL
1mL
500µL
7mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form unitSize
12603427 100 μL Concentrated 0.1mL
12613427 500 μL Concentrated 500µL
12693417 1 mL Concentrated 1mL
12643427 7 mL Prediluted 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12603427 Supplier Epredia Supplier No. RB9055P0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide accurate, reproducible results with the Epredia™ PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody.

Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase delta auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-delta to play a fundamental role in the initiation of cell proliferation.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant human PCNA protein

Molecular Weight: 36kDa

Positive Control: Raji cells, Tonsil, Lymph node, and small intestine

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA (Proliferating Cell Nuclear Antigen)
Applications Western Blot
Classification Polyclonal
Concentration 200µg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant human PCNA protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.