Learn More
PCNA Monoclonal Antibody (PC10 (3F81)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9910-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: The PC10 antibody recognizes the proliferating cell nuclear antigen (PCNA), a 36 kDa protein, also known as polymerase delta auxiliary protein. PC10 antibody reacts with human, mouse, and rat PCNA. The peak expression of PCNA occurs during the S-phase. Applications Reported: The PC10 (a.k.a. 3F81) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining. Applications Tested: The PC10 (a.k.a. 3F81) antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The PC10 antibody has also been tested by immunocytochemistry of methanol-fixed MDCK cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.Specifications
PCNA | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04961, P12004, P17918 | |
Pcna | |
Affinity chromatography | |
RUO | |
18538, 25737, 5111 | |
4° C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
PC10 (3F81) | |
Unconjugated | |
Pcna | |
ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
For Research Use Only.