missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCNA Monoclonal Antibody (24GB2805)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557542
This item is not returnable.
View return policy
Description
PCNA Monoclonal Antibody for Western Blot
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.
Specifications
| PCNA | |
| Monoclonal | |
| Unconjugated | |
| Pcna | |
| ATLD2; cb16; Cyclin; DNA polymerase delta auxiliary protein; etID36690.10; fa28e03; fb36g03; HGCN8729; hypothetical protein LOC515499; MGC8367; Pcna; pcna protein; Pcna/cyclin; PCNAR; POL30; Proliferating cell nuclear antigen; wu:fa28e03; wu:fb36g03; YBR0811; YBR088C | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 18538, 25737, 5111 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB2805 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04961, P12004, P17918 | |
| Pcna | |
| Recombinant protein of human PCNA (aa 1-261). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction