missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA associated factor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32112
This item is not returnable.
View return policy
Description
PCNA associated factor Polyclonal specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).
Specifications
| PCNA associated factor | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunohistochemistry, Immunoprecipitation | |
| NP_055551.1 | |
| PCLAF | |
| The immunogen this antibody was made to, maps to a region between residue 61 to 111 of human PCNA-Associated Factor using the numbering given in entry NP_055551.1 (GeneID 9768). | |
| 100 μL | |
| Cancer | |
| 9768 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction