missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA associated factor Antibody (K11-P1G7*B8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | PCNA associated factor |
|---|---|
| Clone | K11-P1G7*B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18625966
|
Novus Biologicals
NBP2-50536-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680298
|
Novus Biologicals
NBP2-50536 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCNA associated factor Monoclonal antibody specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| PCNA associated factor | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| 9768 | |
| IgG2b κ | |
| Protein A purified |
| K11-P1G7*B8 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor | |
| Peptide Sequence - RKACPLQPDHT -peptide immunogen is identical in mouse (Mus musculus) and rat (Rattus norvegicus). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title