missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA associated factor Antibody (K11-P1G7*B8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50536AF350
This item is not returnable.
View return policy
Description
PCNA associated factor Monoclonal specifically detects PCNA associated factor in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| PCNA associated factor | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PCLAF | |
| Peptide Sequence - RKACPLQPDHT -peptide immunogen is identical in mouse (Mus musculus) and rat (Rattus norvegicus). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry | |
| K11-P1G7*B8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor | |
| Mouse | |
| Protein A purified | |
| Cancer | |
| 9768.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction