missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-456
This item is not returnable.
View return policy
Description
PCNA Polyclonal specifically detects PCNA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PCNA | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Gorilla, Chimpanzee, American alligator, White-tufted-ear marmoset, Crab-eating macaque and American mink. | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:10000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:2000 - 1:10000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 - 1:10000 | |
| P12004 | |
| PCNA | |
| The immunogen this antibody was made to, maps to a region between residues 75-125 of human proliferating cell nuclear antigen using the SwissProt entry P12004 (GeneID 5111). | |
| 100 μL | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Stem Cell Markers | |
| 5111 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction