missing translation for 'onlineSavingsMsg'
Learn More

PCNA Antibody, DyLight 594, Novus Biologicals™

Product Code. 18774832 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18774832 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18774832 Supplier Novus Biologicals Supplier No. NB100456DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCNA Polyclonal antibody specifically detects PCNA in Human, Mouse samples. It is validated for Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCNA
Applications Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residues 75-125 of human proliferating cell nuclear antigen using the SwissProt entry P12004 (GeneID 5111).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5111
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.