missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCNA Antibody (A6-G11-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32725
This item is not returnable.
View return policy
Description
PCNA Monoclonal antibody specifically detects PCNA in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| PCNA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1-2ug/sample, Immunohistochemistry-Paraffin 1:2000 | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Recombinant protein within human PCNA aa 1-261. (Uniprot: P12004) | |
| 100 μL | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 5111 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| A6-G11-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction