missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCMTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584498
This item is not returnable.
View return policy
Description
Immunogen sequence: DLARIYIRRT LRNFINDEMQ AKGIPQRAPP KRKRKRVKQR INTYVF Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
The function remains unknown.
Specifications
| PCMTD1 | |
| Polyclonal | |
| Unconjugated | |
| PCMTD1 | |
| 8430411F12Rik; A030012M09Rik; FLJ10883; LOW QUALITY PROTEIN: protein-L-isoaspartate O-methyltransferase domain-containing protein 1; PCMTD1; protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1; protein-L-isoaspartate O-methyltransferase domain-containing protein 1; RGD1307986 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96MG8 | |
| PCMTD1 | |
| Recombinant protein corresponding to Human PCMTD1. Recombinant protein control fragment (Product #RP-107067). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction