missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102827
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PCM1.
PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
Specifications
| PCM1 | |
| Polyclonal | |
| Unconjugated | |
| PCM1 | |
| 2600002H09Rik; 9430077F19Rik; C030044G17Rik; hPCM-1; mPCM-1; Pcm1; PCM-1; pericentriolar material 1; pericentriolar material 1 protein; pericentriolar material 1, PCM1; pericentriolar material gene 1; PTC4; RET/PCM-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18536, 5108, 81740 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15154, Q9R0L6 | |
| PCM1 | |
| A synthesized peptide derived from human PCM1(Accession Q15154), corresponding to amino acid residues N108-T158. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction