missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCM1 Antibody (CL0206) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52878MFV500
This item is not returnable.
View return policy
Description
PCM1 Monoclonal antibody specifically detects PCM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PCM1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1 | |
| This antibody was developed using a recombinant protein derived from Q15154, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Mitotic Regulators | |
| 5108 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0206 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction