Learn More
PCM1 Antibody (CL0206) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52878JF669
Product Code. 30287964
  • £558.00 / 0.10mL
Description
PCM1 Monoclonal antibody specifically detects PCM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCM1
Monoclonal
Janelia Fluor 669
hPCM-1, pericentriolar material 1, pericentriolar material 1 protein, PTC4PCM-1
This antibody was developed using a recombinant protein derived from Q15154, with the exact immunogen sequence remaining proprietary.
0.1 mL
Mitotic Regulators
5108
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0206
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified