missing translation for 'onlineSavingsMsg'
Learn More

PCL 016, MedChemExpress

Product Code. 16441248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441248 Supplier Medchem Express Supplier No. HYI0660
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PCL 016 is a topical antiviral agent, which inhibits adenovirus replication in rabbit.

PCL 016 is a topical antiviral agent, which inhibits adenovirus replication in rabbit.

Specifications

Chemical Name or Material PCL 016
Physical Form Solid
CAS 98-98-6
Color White to Off-White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C6H5NO2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 100 mg/mL (812.28 mM) ∣H2O : ≥ 50 mg/mL (406.14 mM)
SMILES O=C(O)C1=CC=CC=N1
Molecular Weight (g/mol) 123.11
Formula Weight 123.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.