missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530221
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HeLa, HepG2, Neuro2A, GL261, PC-12. Predicted reactivity: Mouse (93%), Rat (90%), Pig (96%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the phosphoenolpyruvate carboxykinase (GTP) family. The protein is a mitochondrial enzyme that catalyzes the conversion of oxaloacetate to phosphoenolpyruvate in the presence of GTP. A cytosolic form encoded by a different gene has also been characterized and is the key enzyme of gluconeogenesis in the liver. The encoded protein may serve a similar function, although it is constitutively expressed and not modulated by hormones such as glucagon and insulin that regulate the cytosolic form. Alternatively spliced transcript variants have been described.
Specifications
| PCK2 | |
| Polyclonal | |
| Unconjugated | |
| PCK2 | |
| 1810010O14Rik; 9130022B02Rik; HGNC:8725; PCK2; PEP carboxykinase; PEPCK; PEPCK2; PEPCK-M; Phosphoenolpyruvate carboxykinase [GTP], mitochondrial; phosphoenolpyruvate carboxykinase 2 (mitochondrial); phosphoenolpyruvate carboxykinase 2, mitochondrial; phosphoenolpyruvate carboxylase; phosphopyruvate carboxylase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361042, 5106, 74551 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q16822, Q8BH04 | |
| PCK2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human PCK2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction