missing translation for 'onlineSavingsMsg'
Learn More

PCK1 Antibody [DyLight 550], Novus Biologicals™

Product Code. 30735218 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735218 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735218 Supplier Novus Biologicals Supplier No. AF7316L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PCK1 Polyclonal antibody specifically detects PCK1 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCK1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 4.1.1.32, MGC22652, PEPCK1, PEPCKC, PEPCK-CPEP carboxykinase, phosphoenolpyruvate carboxykinase 1 (soluble), phosphoenolpyruvate carboxykinase, cytosolic, phosphoenolpyruvate carboxykinase, cytosolic [GTP], Phosphoenolpyruvate carboxylase, phosphopyruvate carboxylase
Host Species Sheep
Immunogen E. coli-derived recombinant human PCK1, Met1-Ile88, Accession # P35558
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5105
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.