missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCIF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16082-1-AP-20UL
This item is not returnable.
View return policy
Description
PCIF1 may play a role in transcription elongation or in coupling transcription to pre-mRNA processing through its association with the phosphorylated C-terminal domain (CTD) of RNAPII largest subunit.Specifications
| PCIF1 | |
| Polyclonal | |
| Unconjugated | |
| PCIF1 | |
| C20orf67, PCIF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 228866, 63935 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59114, Q9H4Z3 | |
| PCIF1 | |
| PCIF1 Fusion Protein Ag9080 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction