missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCIF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561996
This item is not returnable.
View return policy
Description
Immunogen sequence: SSLVETPPAE NKPRKRQLSE EQPSGNGVKK PKIEIPVTPT GQSVPSSPSI PGTPTLKMWG TSPEDKQQAA LLRPTEVYWD LDIQTNAVIK HR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
PCIF1 may play a role in transcription elongation or in coupling transcription to pre-mRNA processing through its association with the phosphorylated C-terminal domain (CTD) of RNAPII largest subunit.
Specifications
| PCIF1 | |
| Polyclonal | |
| Unconjugated | |
| PCIF1 | |
| 2310022K11Rik; C20orf67; CAPAM; Cap-specific adenosine methyltransferase; F730014I05Rik; hPCIF1; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; Pcif1; PDX-1 C terminus-interacting factor 1; PDX1 C-terminal inhibiting factor 1; phosphorylated CTD interacting factor PCIF1; phosphorylated CTD-interacting factor 1; PPP1R121; protein phosphatase 1, regulatory subunit 121; RGD1310800 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 63935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H4Z3 | |
| PCIF1 | |
| Recombinant protein corresponding to Human PCIF1. Recombinant protein control fragment (Product #RP-101151). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction