missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCID2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551182
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PCID2 protein.
Required for B-cell survival through the regulation of the expression of cell-cycle checkpoint MAD2L1 protein during B cell differentiation. Component of the TREX-2 complex (transcription and export complex 2), composed of at least ENY2, GANP, PCID2, DSS1, and either centrin CETN2 or CETN3 (PubMed:22307388). The TREX-2 complex functions in docking export-competent ribonucleoprotein particles (mRNPs) to the nuclear entrance of the nuclear pore complex (nuclear basket). TREX-2 participates in mRNA export and accurate chromatin positioning in the nucleus by tethering genes to the nuclear periphery. Binds and stabilizes BRCA2 and is thus involved in the control of R-loop-associated DNA damage and transcription-associated genomic instability. R-loop accumulation does not increase in PCID2-depleted cells.
Specifications
| PCID2 | |
| Polyclonal | |
| Unconjugated | |
| PCID2 | |
| A730042J05Rik; CSN12-like protein; F10; fa11f03; HT004; hypothetical protein LOC617943; ik:tdsubc_1e10; im:6909928; PCI domain containing 2; PCI domain-containing protein 2; Pcid2; RGD1307041; RP11-98F14.6; tdsubc_1e10; wu:fa11f03; xx:tdsubc_1e10; zgc:101845 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 234069, 55795 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q5JVF3, Q8BFV2 | |
| PCID2 | |
| Fusion protein of human PCID2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction