missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PCI 34051

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective histone deacetylase 8 (HDAC8) inhibitor (IC50 = 10 nM). Displays >200 fold selectivity over HDAC isoforms 1, 2, 3, 6 and 10. Induces apoptosis in cell lines derived from T cell lymphomas or leukemias.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 296.32 |
| Molecular Weight (g/mol) | 296.326 |
| Chemical Name or Material | PCI 34051 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction