missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PCI 34051
GREENER_CHOICE

Product Code. 30002001
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002001

Brand: Tocris Bioscience™ 4643/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective HDAC8 inhibitor

Potent and selective histone deacetylase 8 (HDAC8) inhibitor (IC50 = 10 nM). Displays >200 fold selectivity over HDAC isoforms 1, 2, 3, 6 and 10. Induces apoptosis in cell lines derived from T cell lymphomas or leukemias.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 950762-95-5
Molecular Formula C17H16N2O3

Specifications

Quantity 10 mg
Formula Weight 296.32
Molecular Weight (g/mol) 296.326
Chemical Name or Material PCI 34051
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.