missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCGF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113770
This item is not returnable.
View return policy
Description
Immunogen sequence: TQPLYNISAN EGTGHFKPLE KKFVRVSGEA TIGHVEKFLR RKMGLDPACQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
Polycomb group (PcG) proteins form multiprotein complexes that regulate expression patterns of developmental and cell proliferation genes. Several members of the PcG contain ring finger domains and are identified as a subclass of RING finger proteins. The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain the RING-type zinc finger conserved domain are generally involved in the ubiquitination pathway of protein degradation. PCGF6 (polycomb group ring finger 6), also known as MBLR or RNF134, is a 350 amino acid nuclear protein that is ubiquitously expressed and contains one RING-type zinc finger. PCGF6 acts as a transcriptional repressor and regulates the level of histone H3K4Me3 by activating SmcY histone demethylase. PCGF6 exists as two alternatively spliced isoforms.
Specifications
| PCGF6 | |
| Polyclonal | |
| Unconjugated | |
| PCGF6 | |
| 4933407A11Rik; AI604840; Mblr; Mel18 and Bmi1-like RING finger; Mel18- and Bmi1-like RING finger protein; Mel18 and Bmi1-like RING finger protein; Pcgf6; polycomb group ring finger 6; polycomb group RING finger protein 6; RING finger protein 134; RNF134 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84108 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BYE7 | |
| PCGF6 | |
| Synthetic peptide directed towards the middle region of human PCGF6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction