missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCGF5, Mouse anti-Human, Clone: 3C10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00084333-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: NDCPRCGNQVHETNPLEMLRLDNTLEEIIFKLVPGLREQELERESEFWKKNKPQENGQDDTSKADKPKVDEEGDENEDDKDYHRSDPQIAICLDCLRNSpecifications
| PCGF5 | |
| Monoclonal | |
| Unconjugated | |
| polycomb group ring finger 5 | |
| MGC16202/RNF159 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3C10 | |
| In 1x PBS, pH 7.4 | |
| NM_032373 | |
| PCGF5 | |
| PCGF5 (NP_115749.2, 51 a.a. ∼ 148 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 84333 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction