missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCGF2, Mouse anti-Human, Clone: 4C10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007703-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: LTLATVPTPSEGTNTSGASECESVSDKAPSPATLPATSSSLPSPATPSHGSPSSHGPPASpecifications
| PCGF2 | |
| Monoclonal | |
| Unconjugated | |
| polycomb group ring finger 2 | |
| MEL-18/MGC10545/RNF110/ZNF144 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 4C10 | |
| In 1x PBS, pH 7.4 | |
| BC004858 | |
| PCGF2 | |
| PCGF2 (AAH04858.1, 236 a.a. ∼ 294 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7703 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction