missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCF11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94620-0.02ml
This item is not returnable.
View return policy
Description
PCF11 Polyclonal antibody specifically detects PCF11 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PCF11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA0824PCF11p homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog (S. cerevisiae), PCF11, cleavage and polyadenylation factor subunit, homolog (S. cerevisiae), pre-mRNA cleavage complex 2 protein Pcf11, Pre-mRNA cleavage complex II protein Pcf11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1256-1555 of human PCF11 (NP_056969.2). PKAYPDNHLSQVDVNELFSKLLKTGILKLSQTDSATTQVSEVTAQPPPEEEEDQNEDQDVPDLTNFTVEELKQRYDSVINRLYTGIQCYSCGMRFTTSQTDVYADHLDWHYRQNRTEKDVSRKVTHRRWYYSLTDWIEFEEIADLEERAKSQFFEKVHEEVVLKTQEAAKEKEFQSVPAGPAGAVESCEICQEQFEQYWDEEEEEWHLKNAIRVDGKIYHPSCYEDYQNTSSFDCTPSPSKTPVENPLNIMLNIVKNELQEPCDSPKVKEERIDTPPACTEESIATPSEIKTENDTVESV | |
| 0.02 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51585 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction