missing translation for 'onlineSavingsMsg'
Learn More

PCF11 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18400987 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400987 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400987 Supplier Novus Biologicals Supplier No. NBP212787AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PCF11 Polyclonal antibody specifically detects PCF11 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCF11
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0824PCF11p homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog (S. cerevisiae), PCF11, cleavage and polyadenylation factor subunit, homolog (S. cerevisiae), pre-mRNA cleavage complex 2 protein Pcf11, Pre-mRNA cleavage complex II protein Pcf11
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1505 and 1555 of human Cleavage and Polyadenylation Factor II Subunit using the numbering given in entry NP_056969.2 (GeneID 51585).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51585
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.