missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCED1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560546
This item is not returnable.
View return policy
Description
Immunogen sequence: LVWNTAMPVG EEVTGGFLPP KLRRQKATFL KNEVVKANFH SATEARKHNF DVLD Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 61%.
Specifications
| PCED1B | |
| Polyclonal | |
| Unconjugated | |
| PCED1B | |
| FAM113B; family with sequence similarity 113, member B; PCED1B; PC-esterase domain containing 1B; PC-esterase domain-containing protein 1B; Protein FAM113B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91523 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96HM7 | |
| PCED1B | |
| Recombinant protein corresponding to Human PCED1B. Recombinant protein control fragment (Product #RP-97862). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction