missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHGC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531382
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji. Predicted reactivity: Mouse (99%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
| PCDHGC4 | |
| Polyclonal | |
| Unconjugated | |
| PCDHGC4 | |
| PCDH-gamma-C4; PCDHGC4; protocadherin gamma subfamily C, 4; protocadherin gamma-C4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56098 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9Y5F7 | |
| PCDHGC4 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human PCDHGC4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction