missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHGC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10857-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.Specifications
| PCDHGC3 | |
| Polyclonal | |
| Unconjugated | |
| PCDHGC3 | |
| PC 43, PC43, PCDH GAMMA C3, PCDH2, PCDHGC3, Protocadherin 2, Protocadherin 43, Protocadherin gamma C3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116782, 5098, 93706 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UN70 | |
| PCDHGC3 | |
| PCDHGC3 Fusion Protein Ag1243 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction