Learn More
Invitrogen™ PCDHGC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113808
Description
Immunogen sequence: IKDNGEPSLS TTATLTVSVT EDSPEARAEF PSGSAPREQK KNLTFYLLLS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
| PCDHGC3 | |
| Polyclonal | |
| Unconjugated | |
| PCDHGC3 | |
| Cadherin like 2; cadherin-like 2; PC 43; PC43; PC-43; PCDH 2; PCDH gamma C3; PCDH2; PCDH-GAMMA-C3; Pcdhgb6; PCDHGC 3; PCDHGC3; protocadherin 2; protocadherin 43; protocadherin gamma subfamily C, 3; protocadherin gamma-C3; Protocadherin2; Protocadherin-2; Protocadherin43; protocadherin-43 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5098 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UN70 | |
| PCDHGC3 | |
| Synthetic peptide directed towards the C terminal region of human PCDHGC3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.