missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PCDHGB2 Monoclonal Antibody (6E9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522246
This item is not returnable.
View return policy
Description
Peptide Sequence: QICGKQPLCV LDFDTVAENP LNIFYIAVIV QDINDNTPLF KQTKINLKIG ESTKPGTTFP LDPALDSDVG PNSLQRYHLN DNEYFDLAEK QTPDGRKYPE LILKHS.
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
| PCDHGB2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y5G2 | |
| PCDHGB2 | |
| PCDHGB2 (NP_061746, 94 a.a. ~ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 6E9 | |
| Unconjugated | |
| PCDHGB2 | |
| 3' RACE product; PCDH-gamma-B2; PCDHGB2; protocadherin gamma B2; protocadherin gamma subfamily B, 2; protocadherin gamma-B2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56103 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction