missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHB16, Mouse anti-Human, Clone: 3H1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00057717-M03.50ug
This item is not returnable.
View return policy
Description
Sequence: MEIGWMHNRRQRQVLVFFVLLSLSGAGAELGSYSVVEETERGSFVANLGKDLGLGLTEMSTRKARIISQGNKQHLQLKAQTGDLLINEKLDREELCGPTEPCILHFQVLMENPLEIFQAELRVIDINDHSPMFTEKEMILKIPENSPLGTEFPLNHALDLDVGSNNVQNYKISPSSHFRVLIHEFRDGRKYPELVLDKELDREEEPQLRLTLTALDGGSPPRSGTAQVRIEVVDINDNAPEFEQPIYKVQIPENSPLGSLVATVSARDLDGGANGKISYTLFQPSEDISKTLEVNPMTGEVRLRKQVDFEMVTSYEVRIKATDGGGLSGKCTLLLQVVDVNDNPPQVTMSALTSPIAENSPEIVVAVFSVSDPDSGNNGKTISSIQEDLPFLLKPSVKNFYTLVTERALDREARAEYNITLTVTDMGTPRLKTEHNITVQISDVNDNAPTFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALRSLDYEALQAFEFRVGATDRGSPALSREALVRVLVLDANDNSPLVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPFLPLPEAAPGQTQANSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSMPEGPFPGRLVDVSGTGTLSQSYQYEVCLTGGSETSEFKFLKPIIPNFSPSpecifications
| PCDHB16 | |
| Monoclonal | |
| Unconjugated | |
| protocadherin beta 16 | |
| KIAA1621/ME1/PCDH-BETA16/PCDH3X/PCDHB8a | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3H1 | |
| In 1x PBS, pH 7.4 | |
| BC036062 | |
| PCDHB16 | |
| PCDHB16 (AAH36062.1, 1 a.a. ∼ 776 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 57717 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction