Learn More
PCDHAC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24314-1-AP-20UL
Description
The PCDHAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PCDHAC1. This antibody recognizes human, mouse antigen. The PCDHAC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PCDHAC1 | |
| 250 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| PCDH ALPHA C1, PCDHAC1, Protocadherin alpha C1 | |
| 20 μL | |
| 56135 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |