missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18075-1-AP-150UL
This item is not returnable.
View return policy
Description
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.Specifications
| PCDHA9 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA9 | |
| KIAA0345, PCDH alpha 9, PCDH ALPHA9, PCDHA9, protocadherin alpha 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192161, 393090, 9752 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91Y11, Q9Y5H5 | |
| Pcdha9 | |
| PCDHA9 Fusion Protein Ag12767 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction