missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18075-1-AP-20UL
This item is not returnable.
View return policy
Description
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.Specifications
| PCDHA9 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA9 | |
| KIAA0345, PCDH alpha 9, PCDH ALPHA9, PCDHA9, protocadherin alpha 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192161, 393090, 9752 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91Y11, Q9Y5H5 | |
| Pcdha9 | |
| PCDHA9 Fusion Protein Ag12767 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering