missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCDHA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10127-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined.Specifications
| PCDHA2 | |
| Polyclonal | |
| Unconjugated | |
| PCDHA2 | |
| PCDH alpha 2, PCDH ALPHA2, PCDHA2, protocadherin alpha 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56146 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y5H9 | |
| PCDHA2 | |
| PCDHA2 Fusion Protein Ag0173 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction